missing translation for 'onlineSavingsMsg'
Learn More

TAF7 Antibody (3G6), Novus Biologicals™

Product Code. 18327329 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327329 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327329 Supplier Novus Biologicals Supplier No. H00006879M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TAF7 Monoclonal antibody specifically detects TAF7 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF7
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3G6
Conjugate Unconjugated
Dilution Western Blot, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH32737
Gene Alias TAF(II)55, TAF2FTBP-associated factor F, TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 55kDa, TAFII-55, TAFII55RNA polymerase II TBP-associated factor subunit F, TATA box binding protein (TBP)-associated factor, RNA polymerase II, F, 55kD, transcription factor IID subunit TAFII55, Transcription initiation factor TFIID 55 kDa subunit, transcription initiation factor TFIID subunit 7, transcription initiation factor TFIID, 55 kDa subunit
Host Species Mouse
Immunogen TAF7 (AAH32737, 130 a.a. ~ 224 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. FIWNHGITLPLKNVRKRRFRKTAKKKYIESPDVEKEVKRLLSTDAEAVSTRWEIIAEDETKEAENQGLDISSPGMSGHRQGHDSLEHDELREIFN
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6879
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.