Learn More
Invitrogen™ TAF6L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116580
Description
Positive Samples: HeLa, HT-29, Jurkat, Mouse thymus, Rat testis Immunogen sequence: MSEREERRFV EIPRESVRLM AESTGLELSD EVAALLAEDV CYRLREATQN SSQFMKHTKR RKLTVEDFNR ALRWSSVEAV CGYGSQEALP MRPAREGELY FPEDREVNLV ELALATNIPK GCAETAVRVH VSYLDGKGNL APQGSVPSAV SSLTDDLLKY YHQVTRAVLG DDPQLMKVAL QDLQTNSKIG ALLPYFVYVV.
In eukaryotic systems, the process of initiating transcription from protein-coding genes requires the presence of RNA polymerase II and a broad family of auxiliary transcription factors. TFIID is a general transcription factor that initiates preinitiation complex assembly through direct interaction with the TATA promoter element. Functioning as a multisubunit complex consisting of a small TATA-binding polypeptide and other TBP-associated factors (TAFs), TFIID mediates promoter responses to various transcriptional activators and repressors. TAF6L, also known as PAF65A, is a 622 amino acid nuclear protein that functions as part of the PCAF (p300/CBP-associated factor) histone acetylase complex.
Specifications
| TAF6L | |
| Polyclonal | |
| Unconjugated | |
| TAF6L | |
| 2810417N14Rik; BB223262; C530024J06Rik; fa05b10; p300/CBP-associated factor (PCAF)-associated factor 65; Paf65a; PAF65-alpha; PCAF-associated factor 65 alpha; PCAF-associated factor 65-alpha; TAF6L; TAF6-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 6L; TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor; TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor, 65kDa; TATA box binding protein associated factor 6 like; TATA-box binding protein associated factor 6 like; wu:fa05b10 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10629, 225895, 309194 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 1.65 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8R2K4, Q9Y6J9 | |
| TAF6L | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human TAF6L (NP_0064641). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.