Learn More
TAF6L Mouse anti-Human, Unconjugated, Clone: 1G5C4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67569-1-IG-20UL
Description
The TAF6L antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TAF6L. This antibody recognizes human, mouse, rat antigen. The TAF6L antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| TAF6L | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, ELISA | |
| 1G5C4 | |
| Unconjugated | |
| 20 μL | |
| 10629 | |
| IgG2b |