missing translation for 'onlineSavingsMsg'
Learn More

TAF6L Antibody, Novus Biologicals™

Product Code. 18327598 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327598 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327598 Supplier Novus Biologicals Supplier No. H00010629B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

TAF6L Polyclonal antibody specifically detects TAF6L in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF6L
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence
Formulation PBS (pH 7.4)
Gene Alias FLJ11136, p300/CBP-associated factor (PCAF)-associated factor 65, PAF65-alpha, PAF65AMGC4288, PCAF-associated factor 65 alpha, PCAF-associated factor 65-alpha, TAF6-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDasubunit 6L, TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associatedfactor, 65 kD, TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associatedfactor, 65kDa
Host Species Mouse
Immunogen TAF6L (AAH08785.1, 1 a.a. - 459 a.a.) full-length human protein. MSEREERRFVEIPRESVRLMAESTGLELSDEVAALLAEDVCYRLREATQNSSQFMKHTKRRKLTVEDFNRALRWSSVEAVCGYGSQEALPMRPAREGELYFPEDREVNLVELALATNIPKGCAETAVRVHVSYLDGKGNLAPQGSVPSAVSSLTDDLLKYYHQVTRAVLGDDPQLMKVALQDLQTNSKIGALLPYFVYVVSGVKSVSHDLEQLHRLLQVARSLFRNPHLCLGPYVRCLVGSVLYCVLEPLAASINPLNDHWTLRDGAALLLSHIFWTHGDLVSGLYQHILLSLQKILADPVRPLCCHYGAVVGLHALGWKAVERVLYPHLSTYWTNLQAVLDDYSVSNAQVKADGHKVYGAILVAVERLLKMKAQAAEPNRGGPGGRGCRRLDDLPWDSLLFQESSSGGGAEPSFGSGLPLPPGGAGPEDPSLSVTLADIYRELYAFFGDSLATRFGTG
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10629
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.