missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF6L Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93341-0.1ml
This item is not returnable.
View return policy
Description
TAF6L Polyclonal antibody specifically detects TAF6L in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| TAF6L | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunoprecipitation 1:500 - 1:1000 | |
| FLJ11136, p300/CBP-associated factor (PCAF)-associated factor 65, PAF65-alpha, PAF65AMGC4288, PCAF-associated factor 65 alpha, PCAF-associated factor 65-alpha, TAF6-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDasubunit 6L, TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associatedfactor, 65 kD, TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associatedfactor, 65kDa | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human TAF6L (NP_006464.1). MSEREERRFVEIPRESVRLMAESTGLELSDEVAALLAEDVCYRLREATQNSSQFMKHTKRRKLTVEDFNRALRWSSVEAVCGYGSQEALPMRPAREGELYFPEDREVNLVELALATNIPKGCAETAVRVHVSYLDGKGNLAPQGSVPSAVSSLTDDLLKYYHQVTRAVLGDDPQLMKVALQDLQTNSKIGALLPYFVYVV | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10629 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction