missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Target | TAF6L |
| Synonym | FLJ11136, p300/CBP-associated factor (PCAF)-associated factor 65, PAF65-alpha, PAF65AMGC4288, PCAF-associated factor 65 alpha, PCAF-associated factor 65-alpha, TAF6-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDasubunit 6L, TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associatedfactor, 65 kD, TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associatedfactor, 65kDa |
| Gene Symbol | TAF6L |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?