missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF6 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | TAF6 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
Description
TAF6 Polyclonal specifically detects TAF6 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation.Specifications
| TAF6 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 6878 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1.0 ug/ml, Chromatin Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DKFZp781E21155, MGC:8964, RNA polymerase II, E, 70/85kD, TAF(II)70, TAF(II)80, TAF2ERNA polymerase II TBP-associated factor subunit E, TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80kDa, TAFII70TAFII-80, TAFII80TAFII-70, TAFII85, Transcription initiation factor TFIID 70 kDa subunit, Transcription initiation factor TFIID 80 kDa subunit, transcription initiation factor TFIID subunit 6 | |
| The immunogen is a synthetic peptide directed towards the C terminal region of human TAF6 (NP_005632). Peptide sequence PSVQPIVKLVSTATTAPPSTAPSGPGSVQKYIVVSLPPTGEGKGGPTSHP | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title