missing translation for 'onlineSavingsMsg'
Learn More

TAF6 Antibody, Novus Biologicals™

Product Code. 18396949 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396949 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18396949 Supplier Novus Biologicals Supplier No. H00006878D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TAF6 Polyclonal antibody specifically detects TAF6 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen TAF6
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_005632.1
Gene Alias DKFZp781E21155, MGC:8964, RNA polymerase II, E, 70/85kD, TAF(II)70, TAF(II)80, TAF2ERNA polymerase II TBP-associated factor subunit E, TAF6 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 80kDa, TAFII70TAFII-80, TAFII80TAFII-70, TAFII85, Transcription initiation factor TFIID 70 kDa subunit, Transcription initiation factor TFIID 80 kDa subunit, transcription initiation factor TFIID subunit 6
Host Species Rabbit
Immunogen TAF6 (NP_005632.1, 1 a.a. - 677 a.a.) full-length human protein. MAEEKKLKLSNTVLPSESMKVVAESMGIAQIQEETCQLLTDEVSYRIKEIAQDALKFMHMGKRQKLTTSDIDYALKLKNVEPLYGFHAQEFIPFRFASGGGRELYFYEEKEVDLSDIINTPLPRVPLDVCLKAHWLSIEGCQPAIPENPPPAPKEQQKAEATEPLKSAKPGQEEDGPLKGKGQGATTADGKGKEKKAPPLLEGAPLRLKPRSIHELSVEQQLYYKEITEACVGSCEAKRAEALQSIATDPGLYQMLPRFSTFISEGVRVNVVQNNLALLIYLMRMVKALMDNPTLYLEKYVHELIPAVMTCIVSRQLCLRPDVDNHWALRDFAARLVAQICKHFSTTTNNIQSRITKTFTKSWVDEKTPWTTRYGSIAGLAELGHDVIKTLILPRLQQEGERIRSVLDGPVLSNIDRIGADHVQSLLLKHCAPVLAKLRPPPDNQDAYRAEFGSLGPLLCSQVVKARAQAALQAQQVNRTTLTITQPRPTLTLSQAPQPGPRTPGLLKVPGSIALPVQTLVSARAAAPPQPSPPPTKFIVMSSSSSAPSTQQVLSLSTSAPGSGSTTTSPVTTTVPSVQPIVKLVSTATTAPPSTAPSGPGSVQKYIVVSLPPTGEGKGGPTSHPSPVPPPASSPSPLSGSALCGGKQEAGDSPPPAPGTPKANGSQPNSGSPQPAP
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6878
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.