missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF5L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19274-1-AP-150UL
This item is not returnable.
View return policy
Description
TAF5L (TAF5-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 5L), also known as PAF65B, is a 589 amino acid protein that localizes to the nucleus and contains six WD repeats. Existing as a component of the PCAF histone acetylase complex, TAF5L interacts with a number of TAFs and, via this interaction, plays a role in histone acetylation and transcription initiation. Defects in the gene encoding TAF5L, which is expressed as multiple alternatively spliced isoforms, are associated with the pathogenesis of type I diabetes.Specifications
| TAF5L | |
| Polyclonal | |
| Unconjugated | |
| TAF5L | |
| PAF65 beta, PAF65B, PCAF associated factor 65 beta, TAF5L | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 102162, 27097, 307927 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75529, Q91WQ5 | |
| Taf5l | |
| TAF5L Fusion Protein Ag5942 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction