missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAF5L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116004
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TAF5L.
TAF5L (TAF5-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 5L), also known as PAF65B, is a 589 amino acid protein that localizes to the nucleus and contains six WD repeats. Existing as a component of the PCAF histone acetylase complex, TAF5L interacts with a number of TAFs and, via this interaction, plays a role in histone acetylation and transcription initiation. Defects in the gene encoding TAF5L, which is expressed as multiple alternatively spliced isoforms, are associated with the pathogenesis of type I diabetes.
Specifications
| TAF5L | |
| Polyclonal | |
| Unconjugated | |
| Taf5l | |
| 1110005N04Rik; AI849020; Paf65b; PAF65-beta; PCAF associated factor 65 beta; PCAF-associated factor 65 beta; TAF5L; TAF5-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 5L; TAF5-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor; TAF5-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor, 65kDa; TATA box binding protein associated factor 5 like; TATA-box binding protein associated factor 5 like | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27097 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O75529 | |
| Taf5l | |
| A synthesized peptide derived from human TAF5L(Accession O75529), corresponding to amino acid residues K303-D353. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction