missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £470.00
Specifications
| Antigen | TAF5 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:20 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18442981
|
Novus Biologicals
NBP1-86970-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18272086
|
Novus Biologicals
NBP1-86970 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TAF5 Polyclonal specifically detects TAF5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| TAF5 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| TAF(II)100, TAF2DTATA box binding protein (TBP)-associated factor 2D, TAF5 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAFII100TAFII-100, TATA box binding protein (TBP)-associated factor, RNA polymerase II, D, 100kD, transcription initiation factor TFIID 100 kD subunit, Transcription initiation factor TFIID 100 kDa subunit, transcription initiation factor TFIID subunit 5 | |
| TAF5 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:20 | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 6877 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TVVSGSASGPAAPGKVGSVAVEDQPDVSAVLSAYNQQGDPTMYEEYYSGLKHFIECSLDCHRAELSQLFYPLFVHMYLELVYNQHENEAKSFFEKFHGDQECYYQDDLRVLSSLTKKEHMKGNETMLDF | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title