Learn More
TAF3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18901-1-AP-20UL
 View more versions of this product
Product Code. 16818744
  • £135.00 / 20µL
Description
Transcription factor TFIID is one of the general factors required for accurate and regulated initiation by RNA polymerase II. TFIID is a multimeric protein complex that plays a central role in mediating promoter responses to various activators and repressors. Required in complex with TBPL2 for the differentiation of myoblasts into myocytes. The complex replaces TFIID at specific promoters at an early stage in the differentiation process. Belongs to the TFIID complex which is composed of TATA binding protein (TBP) and a number of TBP-associated factors (TAFs). Interacts with TAF10 via the histone fold. Interacts with TAF13, TBP, SAP130 and GCN5L2. Interacts with TBPL2.
Specifications
Specifications
TAF3
Polyclonal
Unconjugated
TAF3
TAF(II)140, TAF140, TAF3, TAFII 140, TAFII140, TBP associated factor 3
Rabbit
Antigen Affinity Chromatography
RUO
83860
-20°C
Liquid
Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5VWG9
TAF3
TAF3 Fusion Protein Ag13536
20 μL