missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18901-1-AP-150UL
This item is not returnable.
View return policy
Description
Transcription factor TFIID is one of the general factors required for accurate and regulated initiation by RNA polymerase II. TFIID is a multimeric protein complex that plays a central role in mediating promoter responses to various activators and repressors. Required in complex with TBPL2 for the differentiation of myoblasts into myocytes. The complex replaces TFIID at specific promoters at an early stage in the differentiation process. Belongs to the TFIID complex which is composed of TATA binding protein (TBP) and a number of TBP-associated factors (TAFs). Interacts with TAF10 via the histone fold. Interacts with TAF13, TBP, SAP130 and GCN5L2. Interacts with TBPL2.Specifications
| TAF3 | |
| Polyclonal | |
| Unconjugated | |
| TAF3 | |
| TAF(II)140, TAF140, TAF3, TAFII 140, TAFII140, TBP associated factor 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 83860 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5VWG9 | |
| TAF3 | |
| TAF3 Fusion Protein Ag13536 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction