missing translation for 'onlineSavingsMsg'
Learn More

TAF2 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18433714 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433714 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433714 Supplier Novus Biologicals Supplier No. NBP121372AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TAF2 Polyclonal antibody specifically detects TAF2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF2
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias 150 kDa cofactor of initiator function, CIF150TAFII-150, cofactor of initiator function, 150kD subunit, TAF(II)150, TAF2 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2B, TAFII150RNA polymerase II TBP-associated factor subunit B, TATA box binding protein (TBP)-associated factor, RNA polymerase II, B, 150kD, TBP-associated factor 150 kDa, Transcription initiation factor TFIID 150 kDa subunit, transcription initiation factor TFIID subunit 2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1149 and 1199 of human TBP-associated factor 2 using the numbering given in entry NP_003175.1 (GeneID 6873).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6873
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.