missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF1L Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TAF1L |
|---|---|
| Concentration | 0.51 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16816655
|
Proteintech
55170-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16806655
|
Proteintech
55170-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody is specific to TAF1L.
TAF1L, also named as TAF (II)210, belongs to the TAF1 family. It may act as a functional substitute for TAF1/TAFII250 during male meiosis, when sex chromosomes are transcriptionally silenced.Specifications
| TAF1L | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| Q8IZX4 | |
| 138474 | |
| Peptide | |
| Primary | |
| -20°C | |
| TAF1L |
| 0.51 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| TAF(II)210, TAF1L, TAF2A2, TBP associated factor 1 like, TBP associated factor 210 kDa | |
| TAF1L | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title