missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAF1C Recombinant Rabbit Monoclonal Antibody (3O8T1)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557998
This item is not returnable.
View return policy
Description
Immunogen sequence: PWDPGLTARD LLFRGGCRYR KRPRVVLDVT EQISRFLLDH GDVAFAPLGK LMLENFKLEG AGSRTKKKTV VSVKKLLQDL GGHQPWGCPW AYLSNRQRRF SILGGPILGT SVASHLAELL HEELVLRWEQ LLLDEACTGG ALAWVPG.
Initiation of transcription by RNA polymerase I requires the formation of a complex composed of the TATA-binding protein (TBP) and three TBP-associated factors (TAFs) specific for RNA polymerase I. This complex, known as SL1, binds to the core promoter of ribosomal RNA genes to position the polymerase properly and acts as a channel for regulatory signals. TAF1C is the largest SL1-specific TAF. Initiation of transcription by RNA polymerase I requires the formation of a complex composed of the TATA-binding protein (TBP) and three TBP-associated factors (TAFs) specific for RNA polymerase I. This complex, known as SL1, binds to the core promoter of ribosomal RNA genes to position the polymerase properly and acts as a channel for regulatory signals. This gene encodes the largest SL1-specific TAF. Two transcripts encoding different isoforms have been identified.
Specifications
| TAF1C | |
| Recombinant Monoclonal | |
| 1.04 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q15572, Q6P773 | |
| TAF1C | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 75-221 of human TAF1C (NP_005670.4). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 3O8T1 | |
| Unconjugated | |
| TAF1C | |
| MGC:39976; mTAFI95; RNA polymerase I-specific TBP-associated factor 110 kDa; RNA polymerase I-specific TBP-associated factor 95 kDa; SL1; SL1, 110kD subunit; TAF1C; TAFI110; TAFI95; TATA box binding protein (Tbp)-associated factor, RNA polymerase I, C; TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kD; TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kDa; TATA box binding protein (Tbp)-associated factor, RNA polymerase I, C; transcription factor TAFI95; TATA box binding protein associated factor 1C; TATA box-binding protein-associated factor 1C; TATA box-binding protein-associated factor RNA polymerase I subunit C; TATA box-binding protein-associated factor, RNA polymerase I, subunit C; TATA-box binding protein associated factor, RNA polymerase I subunit C; TATA-box binding protein associated factor, RNA polymerase I, C; TBP-associated factor 1C; TBP-associated factor RNA polymerase I 95 kDa; transcription factor SL1; transcription factor TAFI95; transcription initiation factor SL1/TIF-IB subunit C | |
| Rabbit | |
| Protein A | |
| RUO | |
| 361420, 9013 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction