missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF1C Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13341-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a receptor for inositol 1,4,5-trisphosphate, a second messenger that mediates the release of intracellular calcium. The receptor contains a calcium channel at the C-terminus and the ligand-binding site at the N-terminus. Knockout studies in mice suggest that type 2 and type 3 inositol 1,4,5-trisphosphate receptors play a key role in exocrine secretion underlying energy metabolism and growth.Specifications
| TAF1C | |
| Polyclonal | |
| Unconjugated | |
| TAF1C | |
| SL1, TAF1C, TAFI110, TAFI95, TBP associated factor 1C | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9013 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q15572 | |
| TAF1C | |
| TAF1C Fusion Protein Ag4164 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction