missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF1C Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£161.00 - £383.00
Specifications
| Antigen | TAF1C |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30226459
|
Novus Biologicals
NBP3-33522-20ul |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30232154
|
Novus Biologicals
NBP3-33522-100ul |
100 μL |
£383.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TAF1C Monoclonal antibody specifically detects TAF1C in Human,Rat samples. It is validated for ELISA,Western BlotSpecifications
| TAF1C | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Rat | |
| MGC:39976, RNA polymerase I-specific TBP-associated factor 110 kDa, SL1, 110kD subunit, SL1TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kD, TAFI110TBP-associated factor 1C, TAFI95, TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kDa, TATA box-binding protein-associated factor 1C, TATA box-binding protein-associated factor RNA polymerase I subunit C, transcription factor SL1, Transcription initiation factor SL1/TIF-IB subunit C | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 75-221 of human TAF1C (NP_005670.4).,, Sequence:, PWDPGLTARDLLFRGGCRYRKRPRVVLDVTEQISRFLLDHGDVAFAPLGKLMLENFKLEGAGSRTKKKTVVSVKKLLQDLGGHQPWGCPWAYLSNRQRRFSILGGPILGTSVASHLAELLHEELVLRWEQLLLDEACTGGALAWVPG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 9013 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title