missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF1C Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-12779F
This item is not returnable.
View return policy
Description
TAF1C Polyclonal antibody specifically detects TAF1C in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| TAF1C | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| MGC:39976, RNA polymerase I-specific TBP-associated factor 110 kDa, SL1, 110kD subunit, SL1TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kD, TAFI110TBP-associated factor 1C, TAFI95, TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kDa, TATA box-binding protein-associated factor 1C, TATA box-binding protein-associated factor RNA polymerase I subunit C, transcription factor SL1, Transcription initiation factor SL1/TIF-IB subunit C | |
| The immunogen this antibody was made to, maps to a region between residue 819 and 869 of human TATA Box Binding Protein (TBP)-Associated Factor, RNA Polymerase I, C, 110kDa using the numbering given in entry NP_005670.3 (GeneID 9013). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 9013 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction