missing translation for 'onlineSavingsMsg'
Learn More

TAF1C Antibody (3E6), Novus Biologicals™

Product Code. 18357948 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18357948 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18357948 Supplier Novus Biologicals Supplier No. H00009013M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TAF1C Monoclonal antibody specifically detects TAF1C in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF1C
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3E6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005670
Gene Alias MGC:39976, RNA polymerase I-specific TBP-associated factor 110 kDa, SL1, 110kD subunit, SL1TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kD, TAFI110TBP-associated factor 1C, TAFI95, TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kDa, TATA box-binding protein-associated factor 1C, TATA box-binding protein-associated factor RNA polymerase I subunit C, transcription factor SL1, Transcription initiation factor SL1/TIF-IB subunit C
Host Species Mouse
Immunogen TAF1C (NP_005670.2, 761 a.a. ~ 869 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SLSGHVDPSEDTSSPHSPEWPPADALPLPPTTPPSQELTPDACAQGVPSEQRQMLRDYMAKLPPQRDTPGCATTPPHSQASSVRATRSQQHTPVLSSSQPLRKKPRMGF
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9013
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.