missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAF1B Polyclonal Antibody

Description
Predicted to react with monkey based on sequence homology.
Component of RNA polymerase I core factor complex that acts as a GTF2B/TFIIB-like factor and plays a key role in multiple steps during trancription initiation such as preinitiation complex (PIC) assembly and postpolymerase recruitment events in polymerase I (Pol I) transcription. Binds rDNA promoters and plays a role in Pol I recruitment as a component of the SL1/TIF-IB complex and, possibly, directly through its interaction with RRN3.
Specifications
Specifications
| Antigen | TAF1B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.7 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | TAF1B |
| Gene Accession No. | Q53T94 |
| Gene Alias | 4930408G01Rik; A230108M10Rik; MGC:9349; p63; RAF1B; RAFI63; RNA polymerase I-specific TBP-associated factor 63 kDa; RNA polymerase I-specific TBP-associated factor 68 kDa; SL1; SL1, 63kD subunit; Taf1b; TAFI63; TAFI68; Tafi86; TATA box binding protein (Tbp)-associated factor, RNA polymerase I, B; TATA box binding protein (TBP)-associated factor, RNA polymerase I, B, 63kD; TATA box binding protein (TBP)-associated factor, RNA polymerase I, B, 63kDa; TATA box binding protein associated factor 1B; TATA box-binding protein-associated factor 1B; TATA box-binding protein-associated factor RNA polymerase I subunit B; TATA-box binding protein associated factor, RNA polymerase I subunit B; TATA-box binding protein associated factor, RNA polymerase I, B; TBP-associated factor 1B; TBP-associated factor RNA polymerase I 68; transcription factor TAFI68; transcription initiation factor SL1/TIF-IB subunit B |
| Gene Symbols | TAF1B |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?