missing translation for 'onlineSavingsMsg'
Learn More

TAF1B Antibody, Novus Biologicals™

Product Code. 18370769 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370769 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370769 Supplier Novus Biologicals Supplier No. H00009014B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

TAF1B Polyclonal antibody specifically detects TAF1B in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF1B
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. AAH18137.1
Gene Alias RAF1BTATA box binding protein (TBP)-associated factor, RNA polymerase I, B, 63kD, RAFI63MGC:9349, RNA polymerase I-specific TBP-associated factor 63 kDa, SL1SL1, 63kD subunit, TAFI63TBP-associated factor 1B, TATA box binding protein (TBP)-associated factor, RNA polymerase I, B, 63kDa, TATA box-binding protein-associated factor 1B, TATA box-binding protein-associated factor RNA polymerase I subunit B, Transcription initiation factor SL1/TIF-IB subunit B
Host Species Mouse
Immunogen TAF1B (AAH18137.1, 1 a.a. - 588 a.a.) full-length human protein. MDLEESEEFKERCTQCAAVSWGLTDEGKYYCTSCHNVTERYQEVTNTDLIPNTQIKALNRGLKKKNNTEKGWDWYVCEGFQYILYQQAEALKNLGVGPELKNDVLHNFWKRYLQKSKQAYCKNPVYTTGRKPTVLEDNLSHSDWASEPELLSDVSCPPFLESGAESQSDIHTRKPFPVSKASQSETSVCSGSLDGVEYSQRKEKGIVKMTMPQTLAFCYLSLLWQREAITLSDLLRFVEEDHIPYINAFQHFPEQMKLYGRDRGIFGIESWPDYEDIYKKTIEVGTFLDLPRFPDITEDCYLHPNILCMKYLMEVNLPDEMHSLTCHVVKMTGMGEVDFLTFDPIAKMAKAVKYDVQAVAIIVVVLKLLFLLDDSFEWSLSNLAEKHNEKNKKDKPWFDFRKWYQIMKKAFDEKKQKWEEARAKYLWKSEKPLYYSFVDKPVAYKKREMVVNLQKQFSTLVDSTATAGKKSPSSFQFNWTEEDTDRTCFHGHSLQGVLKEKGQSLLTKNSLYWLSTQKFCRCYCTHVTTYEESNYSLSYQFILNLFSFLLRIKTSLLHEEVSLVEKKLFEKKYSVKRKKSRSKKVRRH
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9014
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.