Learn More
Invitrogen™ TAF172 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596434
Description
Immunogen sequence: LAVQNSSLHD IQHAPKLSAL KQLLLDCGLG NGSTSESGTE SVVAQHRILI FCQLKSMLDI VEHDLLKPHL PSVTYLRLDG SIPPGQRHSI VSRFNNDPSI DVLLLTTHVG GLGLNLTGAD TVVFVEHDWN PMRDLQAMDR AHRIGQKRVV NVYRLITRGT LEEKIMGLQK FKMNIANTVI SQENSSLQSM GTDQLLDLFT LDKDGKAEKA DTSTSGKASM KSILENLSDL WDQEQYDSEY SLENFMHSLK; Positive Samples: HepG2, HeLa, MCF7, Rat testis; Cellular Location: Nucleus.
Initiation of transcription by RNA polymerase II requires the assistance of TATA box-binding protein (TBP) and TBP-associated factors, or TAFs (e.g., TAF2B), in 2 distinct complexes, TFIID and B-TFIID. The TFIID complex is composed of TBP and more than 8 TAFs. However, the majority of TBP is present in the B-TFIID complex, which is composed of TBP and TAFII170, also called TAF172, and has DNA-dependent ATPase activity.
Specifications
| TAF172 | |
| Polyclonal | |
| Unconjugated | |
| BTAF1 | |
| AI414500; AI447930; ATP-dependent helicase BTAF1; BTAF1; BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, (Mot1 homolog, S. cerevisiae); BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170kDa; BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170kDa (Mot1 homolog, S. cerevisiae); B-TFIID TATA-box binding protein associated factor 1; B-TFIID transcription factor-associated 170 kDa subunit; E430027O22Rik; MOT1; Mot1 homolog; RGD1564130; TAF(II)170; TAF170; TAF172; TAF-172; TAFII170; TATA-binding protein-associated factor 172; TBP-associated factor 170; TBP-associated factor 172 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 368042, 9044 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ChIP Assay, ELISA, Immunoprecipitation, Western Blot | |
| 0.57 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O14981 | |
| BTAF1 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.