Learn More
TAF15 Antibody (4D71), mFluor Violet 500 SE, Novus Biologicals™
Product Code. 18430908 Shop All Bio Techne Products
Product Code. 18430908 Supplier Novus Biologicals Supplier No. NBP192685MFV500
Mouse Monoclonal Antibody
TAF15 Monoclonal antibody specifically detects TAF15 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence