missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TAF15 Monoclonal antibody specifically detects TAF15 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Applications | Western Blot, Immunocytochemistry |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence |
| Gene Alias | 68kDa, hTAFII68, Npl3, RBP56TAFII68, RNA-binding protein 56, TAF(II)68, TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2NRBP56/CSMF fusion, TATA box binding protein (TBP)-associated factor, RNA polymerase II, N, 68kD(RNA-binding protein 56), TATA box-binding protein-associated factor 2N (RNA-binding protein 56), TATA-binding protein-associated factor 2N, TBP-associated factor 15,68 kDa TATA-binding protein-associated factor |
| Immunogen | Full length recombinant human TAF15 expressed in and purified from E. coli. [UniProt# Q92804] |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 8148 |
| Content And Storage | Store at 4C in the dark. |
| Form | Purified |
| Isotype | IgG1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?