missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF12 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12353-1-AP-150UL
This item is not returnable.
View return policy
Description
TAF12 is involved in the control of transcription by RNA polymerase II involves the basal transcription machinery which is a collection of proteins. These proteins with RNA polymerase II, assemble into complexes which are modulated by transactivator proteins that bind to cis-regulatory elements located adjacent to the transcription start site. Some modulators interact directly with the basal complex, whereas others may act as bridging proteins linking transactivators to the basal transcription factors. Some of these associated factors are weakly attached while others are tightly associated with TBP in the TFIID complex. Among the latter are the TAF proteins. Different TAFs are predicted to mediate the function of distinct transcriptional activators for a variety of gene promoters and RNA polymerases. TAF12 interacts directly with TBP as well as with TAF2I.Specifications
| TAF12 | |
| Polyclonal | |
| Unconjugated | |
| TAF12 | |
| TAF12, TAF15, TAF2J, TAFII 20/TAFII 15, TAFII20, TAFII20/TAFII15 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 66464, 6883 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q16514, Q8VE65 | |
| Taf12 | |
| TAF12 Fusion Protein Ag3017 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction