missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAF12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546867
This item is not returnable.
View return policy
Description
Peptide sequence: MNQFGPSALI NLSNFSSIKP EPASTPPQGS MANSTAVVKI PGTPGAGGRL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 85%; Rabbit: 100%; Rat: 92%.
TAF12 is involved in the control of transcription by RNA polymerase II involves the basal transcription machinery which is a collection of proteins. These proteins with RNA polymerase II, assemble into complexes which are modulated by transactivator proteins that bind to cis-regulatory elements located adjacent to the transcription start site. Some modulators interact directly with the basal complex, whereas others may act as bridging proteins linking transactivators to the basal transcription factors. Some of these associated factors are weakly attached while others are tightly associated with TBP in the TFIID complex. Among the latter are the TAF proteins. Different TAFs are predicted to mediate the function of distinct transcriptional activators for a variety of gene promoters and RNA polymerases. TAF12 interacts directly with TBP as well as with TAF2I.
Specifications
| TAF12 | |
| Polyclonal | |
| Unconjugated | |
| TAF12 | |
| 20kDa; 2810422D08Rik; AW557038; hypothetical protein LOC614227; TAF12; TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 20 kD; TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 20 kDa; TAF12 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 20kDa; TAF15; TAF2J; TAFII20; TAFII-20; TAFII20/TAFII15; TAFII-20/TAFII-15; TATA box binding protein (TBP)-associated factor, RNA polymerase II, J, 20kD; TATA box binding protein associated factor 12; TATA-box binding protein associated factor 12; TATA-box binding protein associated factor 12; transcription initiation factor TFIID subunit 12; transcription initiation factor TFIID 20 kDa subunits; Transcription initiation factor TFIID 20/15 kDa subunits; transcription initiation factor TFIID subunit 12 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6883 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ChIP Assay, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q16514 | |
| TAF12 | |
| Synthetic peptide directed towards the N-terminal of human TAF12 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction