missing translation for 'onlineSavingsMsg'
Learn More

TAF11 Antibody (3H5), Novus Biologicals™

Product Code. 18335059 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335059 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18335059 Supplier Novus Biologicals Supplier No. H00006882M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TAF11 Monoclonal antibody specifically detects TAF11 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF11
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA
Classification Monoclonal
Clone 3H5
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005634
Gene Alias 28kDa, TAF(II)28, TAF11 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2IMGC:15243, TAFII-28, TAFII28TFIID subunit p30-beta, TATA box binding protein (TBP)-associated factor, RNA polymerase II, I, 28kD, transcription initiation factor TFIID 28 kD subunit, Transcription initiation factor TFIID 28 kDa subunit, transcription initiation factor TFIID subunit 11
Host Species Mouse
Immunogen TAF11 (NP_005634, 158 a.a. ~ 210 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SKVFVGEVVEEALDVCEKWGEMPPLQPKHMREAVRRLKSKGQIPNSKHKKIIF
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6882
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.