missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TAF10 Polyclonal antibody specifically detects TAF10 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | TAF10 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS (pH 7.4) |
| Gene Accession No. | NP_006275.1 |
| Gene Alias | 30kDa, STAF28, TAF10 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2ATAFII-30, TAF2Htranscription initiation factor TFIID 30 kD subunit, TAFII30TAF(II)30, TATA box binding protein (TBP)-associated factor, RNA polymerase II, H, 30kD, Transcription initiation factor TFIID 30 kDa subunit, transcription initiation factor TFIID subunit 10 |
| Host Species | Rabbit |
| Immunogen | TAF10 (NP_006275.1, 1 a.a. - 218 a.a.) full-length human protein. MSCSGSGADPEAAPASAASAPGPAPPVSAPAALPSSTAAENKASPAGTAGGPGAGAAAGGTGPLAARAGEPAERRGAAPVSAGGAAPPEGAISNGVYVLPSAANGDVKPVVSSTPLVDFLMQLEDYTPTIPDAVTGYYLNRAGFEASDPRIIRLISLAAQKFISDIANDALQHCKMKGTASGSSRSKSKDRKYTLTMEDLTPALSEYGINVKKPHYFT |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?