Learn More
TAF1 Recombinant Mouse Monoclonal Antibody, Invitrogen™
Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen 730032
Description
Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and more lot-to-lot consistency.
TAF1 Recombinant Monoclonal antibody specifically detects TAF1 in Human samples. It is validated for Immunoprecipitation.
Specifications
| TAF1 | |
| Monoclonal | |
| Unconjugated | |
| TAF1 | |
| AU015687; B430306D02Rik; BA2R; Ccg1; Ccg-1; CCGS; Cell cycle gene 1 protein; cell cycle, G1 phase defect; complementation of cell cycle block, G1-to-S; DYT3; DYT3/TAF1; KAT4; MRXS33; NSCL2; N-TAF1; OF; P250; RGD1562050; TAF(II)250; TAF1; TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 250kDa; TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, neuron specific isoform; Taf2a; TAFII250; TAFII-250; TATA box binding protein associated factor 1; TATA-box binding protein associated factor 1; TBP-associated factor 250 kDa; transcription factor TFIID p250 polypeptide; transcription initiation factor TFIID 250 kDa subunit; Transcription initiation factor TFIID subunit 1; XDP | |
| 1501-1625 of 1872 of TAF1 Protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunoprecipitation | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide | |
| P21675 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6872 | |
| Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.