missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Chemical Name or Material | TAE Buffer |
| Concentration | 10X |
| Physical Form | Solution |
| Quantity | 5 L |
| Packaging | Upright carton |
| Grade | Molecular Biology, Ultrapure |
Frequently Asked Questions (FAQs)
What are the concentrations of the components of TAE Buffer, 10X Solution, Molecular Biology Grade, Ultrapure (Cat. No. J75904.K8)?
TAE Buffer, 10X Solution, Molecular Biology Grade, Ultrapure contains 400 mM Tris and 0.01 M EDTA in high-purity RO water.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?