missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- TAE running buffer is the most commonly used buffer for DNA agarose gel electrophoresis but is also used for non-denaturing RNA agarose gel electrophoresis.
- Buffer for Electrophoresis.
- Prepare electrophoresis buffers
Specifications
Specifications
| Color | Clear Colorless |
| Physical Form | Solution |
| Quantity | 1 L |
| Formulation | Tris Base: 2M; Acetic Acid : 1M; EDTA : 0.05 M Acetic acid: 1M EDTA: 0.05M |
| MDL Number | MFCD00236357 |
| Synonym | Tris-Borate-EDTA Buffer |
Safety and Handling
Product Identifier
- TAE 50X BUFFER
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- MIXTURE LIST-Contain: acetic acid
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?