missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals TAE (10X), TRIS + acetate + EDTA

Product Code. 15435859
Click to view available options
Quantity:
1 L
2 L
4 L
Unit Size:
1 L
2 L
4 L
This item is not returnable. View return policy

Product Code. 15435859

Brand: Thermo Scientific Chemicals J63677.K3

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TAE (10X), TRIS + acetate + EDTA is a buffer composed of Tris, acetate, and EDTA components and is used in electrophoresis of nucleic acids in agarose and polyacrylamide gels.

TAE is used for DNA agarose gel electrophoresis but is also used for non-denaturing RNA agarose gel electrophoresis, used at various concentrations to study the mobility of DNA in solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • TAE (10X), TRIS + acetate + EDTA allows nucleic acid separation

Application

  • This buffer has been used to prepare agarose and polyacrylamide gels for electrophoresis of nucleic acids
  • It is used as a running and gel preparation buffer, and it is also used for non-denaturing RNA agarose gel electrophoresis
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material TAE (10X)
Physical Form Liquid
Quantity 2 L
MDL Number MFCD00236357
Packaging Plastic bottle
Solubility Information Fully miscible.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.