missing translation for 'onlineSavingsMsg'
Learn More

Tadalafil, MedChemExpress

Product Code. 16474137
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16474137 50 mg 50mg
16427976 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16474137 Supplier Medchem Express Supplier No. HY90009A
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tadalafil (IC-351) is a PDE5 inhibitor with an IC50 value of 1.8 nM.

Tadalafil (IC-351) is a PDE5 inhibitor with an IC50 value of 1.8 nM.

Specifications

Chemical Name or Material Tadalafil
CAS 171596-29-5
Color White
Purity Grade Notes Research
Molecular Formula C22H19N3O4
Quantity 50 mg
Synonym IC-351
Solubility Information DMSO : ≥ 52 mg/mL (133.54 mM)
SMILES O=C([C@@]1([H])CC2=C([C@@H](C3=CC=C(OCO4)C4=C3)N15)NC6=C2C=CC=C6)N(C)CC5=O
Molecular Weight (g/mol) 389.4
Formula Weight 389.4
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.