missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TADA3L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29767
This item is not returnable.
View return policy
Description
TADA3L Polyclonal antibody specifically detects TADA3L in Human samples. It is validated for Immunoprecipitation
Specifications
| TADA3L | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| ADA3, ADA3 homolog, alteration/deficiency in activation 3, epididymis secretory sperm binding protein, FLJ20221, FLJ21329, hADA3, NGG1, STAF54, TADA3L, transcriptional adapter 3, Transcriptional adapter 3-like, transcriptional adaptor 3 | |
| Between 325 and 375 | |
| 100 μg | |
| Cell Cycle and Replication | |
| 10474 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction