missing translation for 'onlineSavingsMsg'
Learn More

TADA3L Antibody (3H3), Novus Biologicals™

Product Code. 18396538 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18396538 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18396538 Supplier Novus Biologicals Supplier No. H00010474M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TADA3L Monoclonal antibody specifically detects TADA3L in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TADA3L
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3H3
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH13433
Gene Alias ADA3, ADA3 homolog, alteration/deficiency in activation 3, epididymis secretory sperm binding protein, FLJ20221, FLJ21329, hADA3, NGG1, STAF54, TADA3L, transcriptional adapter 3, Transcriptional adapter 3-like, transcriptional adaptor 3
Host Species Mouse
Immunogen TADA3L (AAH13433, 1 a.a. ~ 432 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSELKDCPLQFHDFKSVDHLKVCPRYTAVLARSEDDGIGIEELDTLQLELETLLSSASRRLRVLEAETQILTDWQDKKGDRRFLKLGRDHELGAPPKHGKPKKQKLEGKAGHGPGPGPGRPKSKNLQPKIQEYEFTDDPIDVPRIPKNDAPNRFWASVEPYCADITSEEVRTLEELLKPPEDEAEHYKIPPLGKHYSQRWAQEDLLEEQKDGARAAAVADKKKGLMGPLTELDTKDVDALLKKSEAQHEQPEDGCPFGALTQRLLQALVEENIISPMEDSPIPDMSGKESGADGASTSPRNQNKPFSVPHTKSLESRIKEELIAQGLLESEDRPAEDSEDEVLAELRKRQAELKALSAHNRTKKHDLLRLAKEEVSRQELRQRVRMADNEVMDAFRKIMAARQKKRTPTKKEKDQAWKTLKERESILKLLDG
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 10474
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.