missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TADA1L Antibody (PCRP-TADA1-1C9) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14264JF635
This item is not returnable.
View return policy
Description
TADA1L Monoclonal antibody specifically detects TADA1L in Human samples. It is validated for Flow Cytometry,Immunoprecipitation,Western Blot,Protein Array
Specifications
| TADA1L | |
| Monoclonal | |
| Janelia Fluor 635 | |
| ADA1, hADA1, HFI1, SPT3-associated factor 42, SPT3-associated factor 42 (STAF42), STAF42transcriptional adaptor 1 (HFI1 homolog, yeast)-like, TADA1LRP1-9E21.4, transcriptional adapter 1, Transcriptional adapter 1-like protein, transcriptional adaptor 1, transcriptional adaptor 1 (HFI1 homolog, yeast) | |
| Recombinant full-length human TADA1Lprotein (Uniprot: Q96BN2) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunoprecipitation, Western Blot, Protein Array | |
| PCRP-TADA1-1C9 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 117143 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction