missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACI/TNFRSF13B/CVID Antibody (MM0562-9D22) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£171.00 - £444.00
Specifications
| Antigen | TACI/TNFRSF13B/CVID |
|---|---|
| Clone | MM0562-9D22 |
| Concentration | LYOPH |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18056326
|
Novus Biologicals
NBP2-11937-0.025MG |
0.025 mg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18044047
|
Novus Biologicals
NBP2-11937 |
0.1 mg |
£444.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TACI/TNFRSF13B/CVID Monoclonal specifically detects TACI/TNFRSF13B/CVID in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize.Specifications
| TACI/TNFRSF13B/CVID | |
| LYOPH | |
| Unconjugated | |
| Mouse | |
| Human | |
| CD267, CD267 antigen, CVID, CVID2, FLJ39942, MGC133214, TACIMGC39952, TNFRSF14B, Transmembrane activator and CAML interactor, tumor necrosis factor receptor 13B, tumor necrosis factor receptor superfamily member 13B, tumor necrosis factor receptor superfamily, member 13B | |
| TNFRSF13B | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| MM0562-9D22 | |
| Monoclonal | |
| Purified | |
| RUO | |
| O14836 | |
| 23495 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title