missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACI/TNFRSF13B/CVID Antibody (166010) [Janelia Fluor™ 525], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1041JF525
This item is not returnable.
View return policy
Description
TACI/TNFRSF13B/CVID Monoclonal antibody specifically detects TACI/TNFRSF13B/CVID in Mouse samples. It is validated for Flow Cytometry,Western Blot,ELISA Standard (Matched Pair),ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair),CyTOF
Specifications
| TACI/TNFRSF13B/CVID | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CD267, CD267 antigen, CVID, CVID2, FLJ39942, MGC133214, TACIMGC39952, TNFRSF14B, Transmembrane activator and CAML interactor, tumor necrosis factor receptor 13B, tumor necrosis factor receptor superfamily member 13B, tumor necrosis factor receptor superfamily, member 13B | |
| Mouse myeloma cell line NS0-derived recombinant mouse TACI/TNFRSF13B, Phe5-Thr129, Accession # Q9ET35 | |
| 0.5 mL | |
| Apoptosis, B Cell Development and Differentiation Markers, Signal Transduction | |
| 23495 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Western Blot, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF | |
| 166010 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction