missing translation for 'onlineSavingsMsg'
Learn More

Tacedinaline, MedChemExpress

Product Code. 16464057
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464057 50 mg 50mg
16458148 10 mg 10mg
16421336 200 mg 200mg
16407906 100 mg 100mg
16461936 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16464057 Supplier Medchem Express Supplier No. HY50934
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Tacedinaline (N-acetyldinaline) is an inhibitor of the histone deacetylase (HDAC) with IC50s of 0.9, 0.9, 1.2 μM for recombinant HDAC 1, 2 and 3 respectively.

Tacedinaline (N-acetyldinaline) is an inhibitor of the histone deacetylase (HDAC) with IC50s of 0.9, 0.9, 1.2 μM for recombinant HDAC 1, 2 and 3 respectively.

Specifications

Chemical Name or Material Tacedinaline
Physical Form Solid
CAS 112522-64-2
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C15H15N3O2
For Use With (Application) Cancer-programmed cell death
Synonym N-acetyldinaline CI-994 Goe-5549
Solubility Information DMSO : ≥ 58 mg/mL (215.37 mM)
SMILES O=C(NC1=C(N)C=CC=C1)C2=CC=C(NC(C)=O)C=C2
Molecular Weight (g/mol) 269.3
Formula Weight 269.3
Percent Purity 95.77%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.