missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TACE/ADAM17 Cytosolic Domain Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
£448.15
Specifications
Antigen | TACE/ADAM17 |
---|---|
Clone | 136121 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
TACE/ADAM17 Monoclonal specifically detects TACE/ADAM17 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
TACE/ADAM17 | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
P78536 | |
6868 | |
E. coli-derived recombinant human TACE/ADAM17 Asp695-Cys824 (predicted) Accession # P78536 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human TACE/ADAM17 Cytosolic Domain in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) ADAM8, 9, 10, 15, or 33 is observed. |
136121 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ADAM 17, ADAM metallopeptidase domain 17, ADAM metallopeptidase domain 18, ADAM17, ADAM18, CD156b, CD156b antigen, CSVP, disintegrin and metalloproteinase domain-containing protein 17, EC 3.4.24.86, MGC71942, Snake venom-like protease, TACEcSVP, TNF-alpha convertase, TNF-alpha converting enzyme, TNF-alpha-converting enzyme, tumor necrosis factor, alpha, converting enzyme | |
ADAM17 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only