missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACE/ADAM17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15281
This item is not returnable.
View return policy
Description
TACE/ADAM17 Polyclonal specifically detects TACE/ADAM17 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications
| TACE/ADAM17 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7), 40% Glycerol with 0.025% Proclin 300 | |
| ADAM 17, ADAM metallopeptidase domain 17, ADAM metallopeptidase domain 18, ADAM18, CD156B, CD156b antigen, CSVP, disintegrin and metalloproteinase domain-containing protein 17, EC 3.4.24.86, MGC71942, Snake venom-like protease, TACEcSVP, TNF-alpha convertase, TNF-alpha converting enzyme, TNF-alpha-converting enzyme, tumor necrosis factor, alpha, converting enzyme | |
| Rabbit | |
| 93 kDa | |
| 0.1 mL | |
| Apoptosis, Stem Cell Signaling Pathway | |
| 6868 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Knockout Validated | |
| P78536 | |
| ADAM17 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human ADAM17. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction