missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACE/ADAM17 Antibody (5A1), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | TACE/ADAM17 |
|---|---|
| Clone | 5A1 |
| Dilution | ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043337
|
Novus Biologicals
NBP3-26523-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043105
|
Novus Biologicals
NBP3-26523-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TACE/ADAM17 Monoclonal antibody specifically detects TACE/ADAM17 in Human samples. It is validated for ELISA, ImmunocytochemistrySpecifications
| TACE/ADAM17 | |
| ELISA, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ADAM 17, ADAM metallopeptidase domain 17, ADAM metallopeptidase domain 18, ADAM18, CD156B, CD156b antigen, CSVP, disintegrin and metalloproteinase domain-containing protein 17, EC 3.4.24.86, MGC71942, Snake venom-like protease, TACEcSVP, TNF-alpha convertase, TNF-alpha converting enzyme, TNF-alpha-converting enzyme, tumor necrosis factor, alpha, converting enzyme | |
| A synthesized peptide derived from Human TACE/ADAM17 [UniProt P78536] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 5A1 | |
| ELISA, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Apoptosis, Myeloid derived Suppressor Cell, Neurodegeneration, Stem Cell Signaling Pathway | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 6868 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title