missing translation for 'onlineSavingsMsg'
Learn More

TACC3 Antibody (OTI6F1), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18414247 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414247 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414247 Supplier Novus Biologicals Supplier No. NBP274436MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TACC3 Monoclonal antibody specifically detects TACC3 in Human, Monkey, Primate samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen TACC3
Applications Flow Cytometry
Classification Monoclonal
Clone OTI6F1
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias ERIC1ERIC-1, MGC117382, MGC133242, transforming acidic coiled-coil-containing protein 3, transforming, acidic coiled-coil containing protein 3
Host Species Mouse
Immunogen Full length human recombinant protein of human TACC3(NP_006333) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Mitotic Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 10460
Target Species Human, Monkey, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.