missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TACC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599235
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TACC1.
This locus may represent a breast cancer candidate gene. It is located close to FGFR1 on a region of chromosome 8 that is amplified in some breast cancers. Three transcript variants encoding different isoforms have been found for this gene.
Specifications
| TACC1 | |
| Polyclonal | |
| Unconjugated | |
| TACC1 | |
| 4833447E04Rik; AA960202; B230378H13Rik; Ga55; gastric cancer antigen Ga55; KIAA1103; Tacc1; Tacc1a; Taxin-1; transforming acidic coiled-coil containing protein 1; transforming acidic coiled-coil-containing protein 1; transforming, acidic coiled-coil containing protein 1; transforming, acidic coiled-coil containing protein 1a | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 320165, 6867 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75410, Q6Y685 | |
| TACC1 | |
| A synthesized peptide derived from human TACC1(Accession O75410), corresponding to amino acid residues A21-R71. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction