missing translation for 'onlineSavingsMsg'
Learn More

TACA, Tocris Bioscience™

Product Code. 18045742
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18045742

Brand: Tocris Bioscience 0181/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

GABAA agonist; also GABA-T substrate and GABA uptake inhibitor

The trans-isomer of CACA. Potent GABAA agonist, GABA uptake inhibitor and substrate for GABA-T. Also GABAA-ρ agonist.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38090-53-8
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.105
InChI Key FMKJUUQOYOHLTF-OWOJBTEDSA-N
Synonym taca, e-4-amino-2-butenoic acid, 4-aminobut-2-enoic acid, trans-4-aminocrotonic acid, t-4-aminocrotonic acid taca, chembl33086, dsstox_cid_25648, dsstox_rid_81026, dsstox_gsid_45648, 2e-4-aminobut-2-enoic acid
PubChem CID 5310987
IUPAC Name (E)-4-aminobut-2-enoic acid
SMILES C(C=CC(=O)O)N

Specifications

Quantity 10 mg
Formula Weight 101.1
Percent Purity >98%
Chemical Name or Material TACA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado