missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TAC1 C-terminal Peptide Polyclonal Antibody
GREENER_CHOICE

Product Code. 13210709
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13210709 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13210709 Supplier Invitrogen™ Supplier No. PA525766

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse, rat and rabbit based on sequence homology.

Steroid receptors are ligand-dependent, intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate hormone. Glucocorticoids are a family of steroids necessary for the regulation of energy metabolism and the immune and inflammatory responses. These compounds exert their effect through their interaction with the glucocorticoid receptor (GR) and that complex's subsequent association with DNA. All normal mammalian tissues examined to date have been shown to contain glucocorticoid receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAC1 C-terminal Peptide
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene TAC1
Gene Accession No. P20366
Gene Alias C-terminal-flanking peptide; Hs.2563; ne; neurokinin 1; neurokinin 2; neurokinin A; neurokinin alpha; neuromedin L; Neuropeptide gamma; neuropeptide K; NK2; NKA; NKNA; NPK; PPT; preprotachykinin; protachykinin; protachykinin-1; Substance K; substance P; TAC1; TAC2; tachykinin 2; tachykinin precursor 1; tachykinin, precursor 1 (substance K, substance P, neurokinin 1, neurokinin 2, neuromedin L, neurokinin alpha, neuropeptide K, neuropeptide gamma)
Gene Symbols TAC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 102-129 amino acids from the C-terminal region of human TAC1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6863
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.