missing translation for 'onlineSavingsMsg'
Learn More

Palintest™ TAC Test Disc

Product Code. 11988896
Click to view available options
Quantity:
50/Pk.
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 11988896

Brand: Palintest™ CM192

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For use with Palintest™ comparator

The Palintest™ comparator with interchangeable color discs is simple to use. Just add the reagent tablet to the test sample, place in the comparator and match against the appropriate disc.

Specifications

Concentration 0 to 250 mg/L
Packaging 50/Pk.
Quantity 50/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.