missing translation for 'onlineSavingsMsg'
Learn More

Tabimorelin Hemifumarate, TRC

Product Code. 30138687
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138687 1 mg 1mg
30009707 10 mg 10mg
30010052 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138687 Supplier Toronto Research Chemicals Supplier No. TRCT0035051MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 242143-80-2
Molecular Formula C32H40N4O3.1/2C4H4O4
Molecular Weight (g/mol) 586.73
Synonym N-[(2E)-5-Amino-5-methyl-1-oxo-2-hexen-1-yl]-N-methyl-3-(2-naphthalenyl)-D-alanyl-N,Nα-Dimethyl-D-phenylalaninamide Hemifumarate, NN 703 Hemifumarate, NNC 26-0703 Hemifumarate
IUPAC Name (2S,5S)-2-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMILES OC[C@]1(O)OC[C@H](O)C(O)C1O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C6H12O6/c7-2-6(11)5(10)4(9)3(8)1-12-6/h3-5,7-11H,1-2H2/t3-,4?,5?,6-/m0/s1
Formula Weight 180.0634
Chemical Name or Material Tabimorelin Hemifumarate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.