missing translation for 'onlineSavingsMsg'
Learn More

(-)-Tabersonine, TRC

Product Code. 30422510
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422510 25 mg 25mg
30416508 100 mg -
30412396 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30422510 Supplier Toronto Research Chemicals Supplier No. TRCT00390025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4429-63-4
Molecular Formula C21 H24 N2 O2
Molecular Weight (g/mol) 336.43
Synonym (5α,12R,19α)-2,3,6,7-Tetradehydroaspidospermidine-3-carboxylic Acid Methyl Ester, 3a-Ethyl-3a,4,6,11,12,13a-hexahydro-1H-indolizino[8,1-cd]carbazole-5-carboxylic Acid Methyl Ester, Tabersonine, Tabersonin
SMILES CC[C@@]12CC(=C3Nc4ccccc4[C@@]35CCN(CC=C1)[C@@H]25)C(=O)OC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H24N2O2/c1-3-20-9-6-11-23-12-10-21(19(20)23)15-7-4-5-8-16(15)22-17(21)14(13-20)18(24)25-2/h4-9,19,22H,3,10-13H2,1-2H3/t19-,20-,21-/m0/s1
Formula Weight 336.1838
Chemical Name or Material Tabersonine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.